missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™

Catalog Number 10676062
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


33.49 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
ClCCl
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
dichloromethane
ClCCl

CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C

TRUSTED_SUSTAINABILITY
Melting Point -97°C
Color Colorless
Boiling Point 39°C
Quantity 2.5 L
Packaging Plastic-coated Glass Bottle
Physical Form Liquid
Vapor Pressure 350mbar at 20°C
Viscosity 0.43 mPaS at 20°C
Formula Weight 84.93g/mol
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'