missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloromethane, Certified AR for Analysis, Stabilised with Amylene, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic Canister
Plastic-coated Glass Bottle
Description
CH2Cl2, CAS Number-75-09-2, aerothene mm, methane, dichloro, methane dichloride, methylene bichloride, methylene chloride, methylene dichloride, solaesthin, freon 30, narkotil, solmethine, 2.5L, 39 deg.C, CHEBI:15767, Colorless, 84.93g/mol, YMWUJEATGCHHMB-UHFFFAOYSA-N, dichloromethane, -97 deg.C
Specifications
Melting Point | -97°C |
Color | Colorless |
Boiling Point | 39°C |
Quantity | 10 L |
Packaging | Metal Can |
Physical Form | Liquid |
Vapor Pressure | 350mbar at 20°C |
Viscosity | 0.43 mPaS at 20°C |
Formula Weight | 84.93g/mol |
Safety and Handling
Product Identifier
- Dichloromethane
- Warning
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H336-May cause drowsiness or dizziness.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'