missing translation for 'onlineSavingsMsg'
Learn More

Dichloromalononitrile 97%, ACROS Organics™

Brand:  Acros Organics 113450010

Code : 29

Additional Details : CAS Number : 13063-43-9 Weight : 0.05100kg Transport : UN number : 2810 Chem class : 1 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10616492

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Dichloromalononitrile
13063-43-9
97%
NCCCl2CN
MFCD00001841
97%
≥96.0%
C3Cl2N2
134.95

Specifications

Dichloromalononitrile
Clear yellow to light brown
97%
Authentic
≥96.0%
C3Cl2N2
134.95
Glass bottle
02, II, 537
1.3100g/mL
1g
13063-43-9
Liquid
97%
NCCCl2CN
MFCD00001841
1.4310 to 1.4360 (20°C, 589nm)
Videos
SDS