missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloroisocyanuric acid sodium salt monohydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 240.0°C to 250.0°C |
Color | White |
Physical Form | Crystals or Granules |
Quantity | 1 kg |
Assay Percent Range | 96.0 to 104% (on dry substance) (Iodometry) |
Solubility Information | Solubility in water: ca 250g/L (25°C) |
Formula Weight | 237.96 |
Percent Purity | 97% |
Infrared Spectrum | Authentic |
Loss on Drying | 15% max. |
Show More |
Safety and Handling
Product Identifier
- Dichloroisocyanuric acid sodium salt dihydrate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- EUH031-Contact with acids liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'