missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 97%, ACROS Organics™

Brand:  Acros Organics 338100250

Code : 29

Additional Details : CAS Number : 1719-53-5 Weight : 0.07500kg Transport : UN number : 1767 Chem class : 8 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10576641

  • £73.20 / 25g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

Dichlorodiethylsilane
1719-53-5
C4H10Cl2Si
157.12
Diethyldichlorosilane
97%
97%
(CH3CH2)2Si(Cl)2
MFCD00000493

Specifications

Dichlorodiethylsilane
28°C
128.0° to 129.0°C
25g
1719-53-5
97%
C4H10Cl2Si
157.12
Diethyldichlorosilane
1.0500g/mL
1.05
− 96.0°C
97%
Glass bottle
97%
(CH3CH2)2Si(Cl)2
MFCD00000493
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dichlorodiethylsilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS