missing translation for 'onlineSavingsMsg'
Learn More

Dichlorodiethylsilane, 97%, Thermo Scientific Chemicals

Catalog Number p-105332
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1719-53-5
C4H10Cl2Si
157.11
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](Cl)(Cl)CC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1719-53-5
C4H10Cl2Si
157.11
MFCD00000493
BYLOHCRAPOSXLY-UHFFFAOYSA-N
diethyldichlorosilane, silane, dichlorodiethyl, dichloro diethyl silane, diethyldichlorosilicon, diethyl dichlorosilane, diethyidichlorosilane, snpladqptpdumup@, dichlorodiethyl-silan, diethylsiliciumdichlorid, silane,dichlorodiethyl
15589
dichloro(diethyl)silane
CC[Si](Cl)(Cl)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -96.0°C
Density 1.0500g/mL
Boiling Point 128.0°C to 129.0°C
Flash Point 28°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CH2)2Si(Cl)2
Quantity 5 g
Specific Gravity 1.05
Solubility Information Solubility in water: reacts
Formula Weight 157.12
Percent Purity 97%
Chemical Name or Material Dichlorodiethylsilane, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dichlorodiethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'