missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(tri-o-tolylphosphine)palladium(II), 95%, Thermo Scientific Chemicals

Catalog Number 15456655
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
GMLMZLGOMMIIMA-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1
This item is not returnable. View return policy
40691-33-6
C42H42Cl2P2Pd
786.06
MFCD00274659
GMLMZLGOMMIIMA-UHFFFAOYSA-L
palladium,dichlorobis tris 2-methylphenyl phosphine
90656943
palladium;tris(2-methylphenyl)phosphane;dichloride
Cl[Pd++]Cl.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1.CC1=CC=CC=C1P(C1=C(C)C=CC=C1)C1=C(C)C=CC=C1

Dichlorobis(tri-o-tolylphosphine)palladium(II) is used in reaction of tributyltin enolates, prepared in situ from tributyltin methoxide and enol acetates, with aryl bromides, coupling reaction of aryl bromides with vinylic acetates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 1 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in ethanol,benzene,and ethereal solvents.
Formula Weight 786.08
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dichlorobis (tri-o-tolylphosphine)palladium (II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'