missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(di-tert-butylphenylphosphine)palladium(II), Pd 17.1%, Thermo Scientific Chemicals

Catalog Number p-4408401
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
34409-44-4
C28H46Cl2P2Pd
621.94
MFCD09953450
DFKFLLBQLYGVIH-UHFFFAOYSA-L
dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride
56649816
ditert-butyl(phenyl)phosphane;dichloropalladium
Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C
This item is not returnable. View return policy
34409-44-4
C28H46Cl2P2Pd
621.94
MFCD09953450
DFKFLLBQLYGVIH-UHFFFAOYSA-L
dichlorobis di-tert-butylphenylphosphine palladium ii, bis di-t-butyl phenyl phosphine palladium dichloride, bis di-tert-butyl phenyl phosphane ; palladium chloride
56649816
ditert-butyl(phenyl)phosphane;dichloropalladium
Cl[Pd++]Cl.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C.CC(C)(C)P(C1=CC=CC=C1)C(C)(C)C

Dichlorobis(di-tert-butylphenylphosphine)palladium(II) is used as an effective catalyst for cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point (decomposition)
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water and polar organic solvents.
Formula Weight 621.95
Physical Form Solid
Chemical Name or Material Dichlorobis(di-tert-butylphenylphosphine)palladium(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'