missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II), 97%, Thermo Scientific™

Catalog Number p-4228587
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1092372-91-2
C20H48Cl2N2P2Ru
550.54
MFCD07782003
FUALRLYVUZSWKT-UHFFFAOYSA-L
dichlorobis 2-di-tert-butylphosphino ethylamine ruthenium ii, dichlorobis 2-di-t-butylphosphino ethylamine ruthenium ii
71310624
2-ditert-butylphosphanylethanamine;dichlororuthenium
Cl[Ru]Cl.CC(C)(C)P(CCN)C(C)(C)C.CC(C)(C)P(CCN)C(C)(C)C
This item is not returnable. View return policy
1092372-91-2
C20H48Cl2N2P2Ru
550.54
MFCD07782003
FUALRLYVUZSWKT-UHFFFAOYSA-L
dichlorobis 2-di-tert-butylphosphino ethylamine ruthenium ii, dichlorobis 2-di-t-butylphosphino ethylamine ruthenium ii
71310624
2-ditert-butylphosphanylethanamine;dichlororuthenium
Cl[Ru]Cl.CC(C)(C)P(CCN)C(C)(C)C.CC(C)(C)P(CCN)C(C)(C)C
Melting Point 247°C to 263°C
Color Brown to Yellow
Elemental Analysis 41.9 to 45.4% C, 4.9 to 5.3% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 550.54
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
compliance-icons
Product Identifier
  • Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'