missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II), 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 247°C to 263°C |
Color | Brown to Yellow |
Elemental Analysis | 41.9 to 45.4% C, 4.9 to 5.3% N |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 550.54 |
Percent Purity | 97% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II) |
Safety and Handling
Product Identifier
- Dichlorobis(2-(di-tert-butylphosphino)ethylamine)ruthenium(II)
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'