missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetyl chloride, 97%, Thermo Scientific™

Catalog Number p-104654
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
79-36-7
C2HCl3O
147.39
MFCD00000840
FBCCMZVIWNDFMO-UHFFFAOYSA-N
dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
6593
CHEBI:34688
2,2-dichloroacetyl chloride
C(C(=O)Cl)(Cl)Cl
This item is not returnable. View return policy
79-36-7
C2HCl3O
147.39
MFCD00000840
FBCCMZVIWNDFMO-UHFFFAOYSA-N
dichloroacetyl chloride, dichloracetyl chloride, acetyl chloride, dichloro, dichloroethanoyl chloride, chlorure de dichloracetyle, dichloroacetylchloride, dichloroacetic acid chloride, alpha,alpha-dichloroacetyl chloride, chcl2cocl, chlorid kyseliny dichloroctove
6593
CHEBI:34688
2,2-dichloroacetyl chloride
C(C(=O)Cl)(Cl)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.5300g/mL
Boiling Point 107.0°C to 108.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Cl2CHCOCl
Refractive Index 1.4600 to 1.4620
Quantity 100 mL
Beilstein 02, 204
Merck Index 15, 3062
Specific Gravity 1.53
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in ether,decomposes in alcohol
Formula Weight 147.39
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dichloroacetyl chloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloroacetyl chloride
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'