missing translation for 'onlineSavingsMsg'
Learn More

Dichloroacetic anhydride, 98%, ACROS Organics™

Brand:  Acros Organics 165120500

Code : 29

Additional Details : CAS Number : 4124-30-5 Weight : 0.10000kg Transport : UN number : 2923 Chem class : 1a Pack group : II


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10562951

  • £178.10 / 50g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

Dichloroacetic anhydride
4124-30-5
98%
(Cl2CHCO)2O
MFCD00000842
98%
98%
C4H2Cl4O3
239.87

Specifications

Dichloroacetic anhydride
1.57
Clear colorless
50g
4124-30-5
Authentic
1.4825 to 1.4845
98%
C4H2Cl4O3
239.87
02, 204
>112°C
214.0° to 216.0°C
29.0°C
98%
1.5700g/mL
Glass bottle
Liquid After Melting
98%
(Cl2CHCO)2O
MFCD00000842
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dichloroacetic anhydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS