missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dichloro-o-xylene, 98%, Thermo Scientific Chemicals

Catalog Number 10695392
Click to view available options
100g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 612-12-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Beige to White
Boiling Point 239°C to 241°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8Cl2
Linear Formula C6H4(CH2Cl)2
MDL Number MFCD00000903
Quantity 100 g
Beilstein 05, 364
Synonym 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid
Solubility Information Solubility in water: hydrolysis
InChI Key FMGGHNGKHRCJLL-UHFFFAOYSA-N
SMILES ClCC1=CC=CC=C1CCl
IUPAC Name 1,2-bis(chloromethyl)benzene
Molecular Weight (g/mol) 175.05
PubChem CID 11919
Formula Weight 175.06
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α, α'-Dichloro-o-xylene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a'-Dichloro-o-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'