missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dichloro(norbornadiene)platinum(II), Pt 54.4%

Brand:  Alfa Aesar™ 044035.03

Code : NEW

Additional Details : CAS Number : 12152-26-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15481947

  • £139.92 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Dichloro (norbornadiene)platinum (II)
358.13
PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
12152-26-0
MFCD00213423

Specifications

Dichloro (norbornadiene)platinum (II)
Cream
1g
Crystalline
MFCD00213423
Soluble in acetic acid
Pt 54.4%
≈200°C (dec.)
12152-26-0
358.13
PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
Videos
SDS