missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dichloro(norbornadiene)platinum(II), Pt 54.4%

Brand:  Alfa Aesar™ 044035.06

Code : NEW

Additional Details : CAS Number : 12152-26-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15491947

  • £611.62 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Dichloro(norbornadiene)platinum(II)
358.13
PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
12152-26-0
MFCD00213423

Specifications

Dichloro(norbornadiene)platinum(II)
Cream
5g
Crystalline
MFCD00213423
Soluble in acetic acid
Pt 54.4%
≈200°C (dec.)
12152-26-0
358.13
PtCl2(nbd); (Bicyclo[2.2.1]hepta-2,5-diene)dichloroplatinum(II)
Videos
SDS