missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(ethylenediamine)platinum(II), Pt 59.7% min, Thermo Scientific Chemicals

Catalog Number p-7048423
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
14096-51-6
C2H8Cl2N2Pt
326.08
MFCD00000011
MAIWCYPHGIXXQM-UHFFFAOYSA-L
dichloroplatinumbis(ylium); ethane-1,2-diamine
Cl[Pt++]Cl.NCCN
This item is not returnable. View return policy
14096-51-6
C2H8Cl2N2Pt
326.08
MFCD00000011
MAIWCYPHGIXXQM-UHFFFAOYSA-L
dichloroplatinumbis(ylium); ethane-1,2-diamine
Cl[Pt++]Cl.NCCN

It is also used as a reactant for Electro reduction reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 0.25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 326.1
Concentration or Composition (by Analyte or Components) Pt 59.7% min.
Physical Form Crystalline Powder
Chemical Name or Material Dichloro(ethylenediamine)platinum(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'