missing translation for 'onlineSavingsMsg'
Learn More

Dichloro(ethylenediamine)palladium(II), Pd 44.8%, Thermo Scientific Chemicals

Catalog Number p-7048422
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15020-99-2
C2H8Cl2N2Pd
237.42
MFCD00044991
CAYKJANQVKIYPJ-UHFFFAOYSA-L
dichloro 1,2-diaminoethane palladium, dichloro ethylenediamine palladium ii, dichloro 1,2-diaminoethane ;palladium, dichloro ethylenediamine palladate ii, ethylenediamine palladium ii chloride, ethylenediamine; palladium chloride, palladium ii chloride ethylenediamine complex, dichloro ethylenediamine palladium ii , pd
11139157
Cl[Pd]Cl.NCCN
This item is not returnable. View return policy
15020-99-2
C2H8Cl2N2Pd
237.42
MFCD00044991
CAYKJANQVKIYPJ-UHFFFAOYSA-L
dichloro 1,2-diaminoethane palladium, dichloro ethylenediamine palladium ii, dichloro 1,2-diaminoethane ;palladium, dichloro ethylenediamine palladate ii, ethylenediamine palladium ii chloride, ethylenediamine; palladium chloride, palladium ii chloride ethylenediamine complex, dichloro ethylenediamine palladium ii , pd
11139157
Cl[Pd]Cl.NCCN

Catalyst for C-C coupling reactions like Sonogashira coupling. Formation of host-guest complexes. Synthesis of mixed-metal, mixed-pyrimidine self-assembling metallacalix[n]arenes; ring-in-ring complex; binding of metal with glycose phosphates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Very low solubility in water.
IUPAC Name dichloropalladium; ethane-1,2-diamine
Formula Weight 237.41
Concentration or Composition (by Analyte or Components) Pd 44.8%
Physical Form Crystalline
Chemical Name or Material Dichloro(ethylenediamine)palladium(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'