missing translation for 'onlineSavingsMsg'
Learn More

Dichloro[bis(dicyclohexylphosphino)propane]palladium(II), Thermo Scientific Chemicals

Catalog Number 15467305
Click to view available options
Quantity:
1 g

 

Dichloro[bis(dicyclohexylphosphino)propane]palladium(II) is used as a catalyst in coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 1041005-52-0
Color White
Molecular Formula C27H52Cl2P2Pd+2
MDL Number MFCD14155689
Quantity 1 g
Synonym dichloro bis dicyclohexylphosphino propane palladium ii, pdcl2 dcypp, bis dicyclohexylphosphino propane dibromopalladium ii
Solubility Information Slightly soluble in dichloromethane and chloroform.
InChI Key JLEVAMQVDDHWOI-UHFFFAOYSA-N
SMILES C1CCC(CC1)[PH+](CCC[PH+](C2CCCCC2)C3CCCCC3)C4CCCCC4.Cl[Pd]Cl
IUPAC Name dichloropalladium;dicyclohexyl(3-dicyclohexylphosphaniumylpropyl)phosphanium
Molecular Weight (g/mol) 615.981
PubChem CID 58567957
Formula Weight 613.97
Physical Form Crystalline Solid
Chemical Name or Material Dichloro[bis(dicyclohexylphosphino)propane]palladium(II)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'