missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II), Thermo Scientific Chemicals
25.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
Description
Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II) is used as an effective catalyst for Kumada coupling and Suzuki reactions. It also catalyzes other reactions to convert enol ethers, dithioacetals and vinyl sulfides to olefins. Further, it is used as a catalyst for preparation of block-copolythiophenes and solid state metathesis polycondensation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 213°C (decomposition) |
Quantity | 2 g |
Solubility Information | Insoluble in water. |
Formula Weight | 542.03 |
Physical Form | Powder |
Chemical Name or Material | Dichloro[bis(1,3-diphenylphosphino)propane]nickel(II) |
Safety and Handling
Product Identifier
- Dichloro¢bis(1,3-diphenylphosphino)propane!nickel(II)
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'