missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with acetone, Pd 13.0-15.0%

Coupling reactions, carbonylation

Brand:  Alfa Aesar™ 044972.06

Code : NEW

Additional Details : CAS Number : 851232-71-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15474925

  • £106.40 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

Dichloro[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
Complex with acetone
787.81
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride; PdCl2(dppf).(CH3)2CO
851232-71-8
C34H28Cl2FeP2Pd·(CH3)2CO
MFCD00792899

Specifications

Dichloro[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
851232-71-8
C34H28Cl2FeP2Pd·(CH3)2CO
MFCD00792899
Orange-red
Pd 13.0 to 15.0%
Complex with acetone
Powder
787.81
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride; PdCl2(dppf).(CH3)2CO
280°C
5g
Videos
SDS