missing translation for 'onlineSavingsMsg'
Learn More

Dicalite™ speed plus, Diatomaceous Filter-aid, Thermo Scientific Chemicals

Catalog Number 10714261
29.75 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY
Physical Form Powder
Density <26.0 lb./ft3 (wet density)
Color White
Quantity 1 kg
pH 9 to 10.5 (10% slurry)
CAS 68855-54-9
Particle Size Retained on 40 Mesh sieve: <0.90 wt.%, retained on 140 Mesh sieve: <10.0 wt.%
Packaging Plastic Bottle
Permeability 85 to 115 (permeability flow ratio)
Molecular Formula CNa2O3
MDL Number MFCD00146688
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.988
PubChem CID 10340
ChEBI CHEBI:29377
Chemical Name or Material Dicalite™ speed plus
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'