missing translation for 'onlineSavingsMsg'
Learn More

Dibutyryl-cAMP, sodium salt, Tocris Bioscience™

Cell-permeable cAMP analog

Brand:  Tocris (Bio-Techne) 1141/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13468723

  • £110.25 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell-permeable analog of cAMP; activates cAMP-dependent protein kinases. Promotes differentiation of dopaminergic neurons (iDA) from hPSCs in combination with other reagents.
Specifications

Chemical Identifiers

16980-89-5
494.397
bucladesine
[(4aR,6R,7R,7aR)-6-[6-(butanoylamino)purin-9-yl]-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-yl] butanoate;molecular hydrogen;sodium
C18H26N5NaO8P
KSSDVZRQZQECKZ-VMZSDVDQSA-N
131673831
[HH].CCCC(=O)NC1=NC=NC2=C1N=CN2C3C(C4C(O3)COP(=O)(O4)O)OC(=O)CCC.[Na]

Specifications

10mg
16980-89-5
bucladesine
[HH].CCCC(=O)NC1=NC=NC2=C1N=CN2C3C(C4C(O3)COP(=O)(O4)O)OC(=O)CCC.[Na]
494.397
491.37
Dibutyryl-cAMP, sodium salt
C18H26N5NaO8P
KSSDVZRQZQECKZ-VMZSDVDQSA-N
[(4aR,6R,7R,7aR)-6-[6-(butanoylamino)purin-9-yl]-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-yl] butanoate;molecular hydrogen;sodium
131673831
>98%
Videos
Documents