missing translation for 'onlineSavingsMsg'
Learn More

Dibutyltin dichloride, 97%, ACROS Organics™

Brand:  Acros Organics 194870010

Code : 29

Additional Details : CAS Number : 683-18-1 Weight : 1.05000kg Transport : UN number : 2928 Chem class : 1a Pack group : II


Quantity 1kg
Packaging Glass bottle

 View more versions of this product

Product Code. 10543561

  • £233.60 / 1kg


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

Dibutyltin dichloride
97%
[CH3(CH2)3]2SnCl2
MFCD00000518
97%
C8H18Cl2Sn
303.83
Dibutyldichlorotin

Specifications

Dibutyltin dichloride
Authentic
Crystalline Low Melting Mass
97%
[CH3(CH2)3]2SnCl2
MFCD00000518
1.4000g/mL
White to beige
135.0°C (10.0 mmHg)
112°C
683-18-1
Glass bottle
97%
C8H18Cl2Sn
303.83
01,213; 01,1294; 05,622
Dibutyldichlorotin
39.0° to 41.0°C
1kg
1.4
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dibutyltin dichloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1

Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS