missing translation for 'onlineSavingsMsg'
Learn More

Dibutylboron trifluoromethanesulfonate, 1M solution in dichloromethane, AcroSeal™, Thermo Scientific™

Catalog Number p-6827170
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
60669-69-4, 60-29-7
C9H18BF3O3S
274.11
MFCD00009669
FAVAVMFXAKZTMV-UHFFFAOYSA-N
dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane
2724243
dibutylboranyl trifluoromethanesulfonate
CCCCB(CCCC)OS(=O)(=O)C(F)(F)F
This item is not returnable. View return policy
60669-69-4, 60-29-7
C9H18BF3O3S
274.11
MFCD00009669
FAVAVMFXAKZTMV-UHFFFAOYSA-N
dibutylboron trifluoromethanesulfonate, dibutylboryl trifluoromethanesulfonate solution, nbu2botf, bu2botf, dibutylboron triflate, n-bu 2botf, dibutylboryl trifluoromethanesulfonate, dibutylboron triflate solution, trifluoromethylsulfonyloxy dibutylborane, dibutyl trifluoromethyl sulfonyl oxy borane
2724243
dibutylboranyl trifluoromethanesulfonate
CCCCB(CCCC)OS(=O)(=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Orange to Yellow
Density 0.8150g/mL
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CF3SO3B[(CH2)3CH3]2
Quantity 10 g
Fieser 07,91; 09,140; 16,109
Formula Weight 274.11
Percent Purity 98%
Physical Form Solution
Chemical Name or Material Dibutylboron trifluoromethanesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Dibutylboron trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'