missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl succinate, 99%, Thermo Scientific Chemicals

Catalog Number 15289826
Click to view available options
Quantity:
5 mL
25 mL
100 mL
This item is not returnable. View return policy
141-03-7
C12H22O4
230.3
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
8830
CHEBI:82263
dibutyl butanedioate
CCCCOC(=O)CCC(=O)OCCCC
This item is not returnable. View return policy
141-03-7
C12H22O4
230.3
YUXIBTJKHLUKBD-UHFFFAOYSA-N
dibutyl succinate, tabutrex, succinic acid di-n-butyl ester, di-n-butyl succinate, tabatrex, dibutylsuccinate, butyl butanedioate, butanedioic acid, dibutyl ester, succinic acid dibutyl ester, dnbs
8830
CHEBI:82263
dibutyl butanedioate
CCCCOC(=O)CCC(=O)OCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -29.0°C
Density 0.9800g/mL
Boiling Point 274.0°C
Flash Point 144°C
Assay Percent Range 98.5% min. (GC)
Linear Formula C4H9O2CH2CH2CO2C4H9
Quantity 5 mL
Specific Gravity 0.98
Solubility Information Solubility in water: slightly.
Formula Weight 230.3
Percent Purity 99%
Chemical Name or Material Dibutyl succinate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'