missing translation for 'onlineSavingsMsg'
Learn More

Dibutyl phosphate, 97%, Thermo Scientific Chemicals

Catalog Number p-103408
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
107-66-4
C8H19O4P
210.21
MFCD00015245
JYFHYPJRHGVZDY-UHFFFAOYSA-N
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
7881
dibutyl hydrogen phosphate
CCCCOP(O)(=O)OCCCC
This item is not returnable. View return policy
107-66-4
C8H19O4P
210.21
MFCD00015245
JYFHYPJRHGVZDY-UHFFFAOYSA-N
dibutyl phosphate, di-n-butyl phosphate, phosphoric acid, dibutyl ester, dibutyl acid phosphate, di-n-butylphosphoric acid, dp 4 coupling agent, di-n-butyl hydrogen phosphate, unii-0072nn74tn, phosphoric acid di-n-butyl ester, phosphoric acid dibutyl ester
7881
dibutyl hydrogen phosphate
CCCCOP(O)(=O)OCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 259 to 267mg KOH/g
Melting Point -13.0°C
Density 1.0580g/mL
Boiling Point 250.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3(CH2)3O)2P(O)OH
Refractive Index 1.4270 to 1.4290
Quantity 250 g
Specific Gravity 1.058
Solubility Information Solubility in water: 17.2g/L
Viscosity 75 mPa.s (20°C)
Formula Weight 210.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dibutyl phosphate, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dibutyl phosphate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'