missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dibromo-p-xylene, 97%, Thermo Scientific Chemicals

Catalog Number 10520551
41.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
623-24-5
C8H8Br2
263.95
MFCD00000182
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
69335
1,4-bis(bromomethyl)benzene
C1=CC(=CC=C1CBr)CBr
623-24-5
C8H8Br2
263.95
MFCD00000182
RBZMSGOBSOCYHR-UHFFFAOYSA-N
1,4-bis bromomethyl benzene, p-xylylene dibromide, alpha,alpha'-dibromo-p-xylene, benzene, 1,4-bis bromomethyl, p-xylylene bromide, p-bromomethyl benzene, 1,4-dibromomethylbenzene, p-bis bromomethyl benzene, 1,4-bis-bromomethyl-benzene, ccris 1775
69335
1,4-bis(bromomethyl)benzene
C1=CC(=CC=C1CBr)CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 142.0°C to 147.0°C
Color White to Beige
Density 2.0120g/mL
Boiling Point 245.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CH2Br)2
Quantity 25 g
Beilstein 05, 385
Specific Gravity 2.012
Solubility Information Solubility in water: soluble,hydrolyses
Formula Weight 263.95
Percent Purity 97%
Physical Form Crystalline Powder and Coarse Crystals
Chemical Name or Material α,α′-Dibromo-p-xylene, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'