Alfa Aesar™ Dibromo[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 12.9%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 044980.03

UNSPSC: 12141733

Code: NEW

Additional Details:
Additional Details: CAS Number: 124268-93-5 Weight: 0.00100kg


Quantity 1g

Product Code. 15436665

Quantity Price
1 £ 84.59 / 1g
Estimated Shipment date
from Supplier 10-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Dibromo[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
CAS 124268-93-5
Molecular Formula C34H28Br2FeP2Pd
Formula Weight 820.62
MDL Number MFCD09953448
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide; PdBr2(dppf)

Specification

Chemical Name or Material Dibromo[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
CAS 124268-93-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Molecular Formula C34H28Br2FeP2Pd
Formula Weight 820.62
MDL Number MFCD09953448
Synonym [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide; PdBr2(dppf)
Concentration Pd 12.9%
Quantity 1g
Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.