missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dibromo[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd 12.9%

Brand:  Alfa Aesar™ 044980.06

UNSPSC : 12141733

Code : NEW

Additional Details : CAS Number : 124268-93-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15446665

  • £306.23 / 5g


Estimated Shipment date
from Supplier 06-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Dibromo[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
C34H28Br2FeP2Pd
MFCD09953448
124268-93-5
820.62
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide; PdBr2(dppf)

Specifications

Dibromo[1,1'-bis (diphenylphosphino)ferrocene]palladium (II)
Solid
820.62
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) bromide; PdBr2(dppf)
5g
124268-93-5
C34H28Br2FeP2Pd
MFCD09953448
Pd 12.9%
Documentation

Technical Documents