missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dibenzylmercury

Brand:  Alfa Aesar™ 037121.06

Code : NEW

Additional Details : CAS Number : 780-24-5 Weight : 0.00500kg Transport : UN number : 2025 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11314297

  • £73.99 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Dibenzylmercury
382.86
UN2025
780-24-5
MFCD00014429

Specifications

Dibenzylmercury
5g
Air Sensitive
382.86
UN2025
108° to 110°C
780-24-5
Crystalline
MFCD00014429
Videos
SDS