missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl N,N-diisopropylphosphoramidite, 90+%, Thermo Scientific Chemicals

Catalog Number p-4813454
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
108549-23-1
C20H28NO2P
345.42
ANPWLBTUUNFQIO-UHFFFAOYSA-N
dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
196621
N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2
This item is not returnable. View return policy
108549-23-1
C20H28NO2P
345.42
ANPWLBTUUNFQIO-UHFFFAOYSA-N
dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
196621
N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.0280g/mL
Flash Point 69°C
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.028
Formula Weight 345.42
Percent Purity 90+%
Chemical Name or Material Dibenzyl N, N-diisopropylphosphoramidite
compliance-icons
Product Identifier
  • Dibenzyl N,N-diisopropylphosphoramidite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'