missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Catalog Number p-103279
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2449-05-0
C16H14N2O4
298.29
MFCD00016737
IRJKSAIGIYODAN-ISLYRVAYSA-N
dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
5387121
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
2449-05-0
C16H14N2O4
298.29
MFCD00016737
IRJKSAIGIYODAN-ISLYRVAYSA-N
dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
5387121
benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen), Radical initiators
TRUSTED_SUSTAINABILITY
Melting Point 44°C to 52°C
Color Yellow to Orange
Flash Point >230°C
Loss on Drying 0.75% max.
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 5 g
Fieser 15,111
Solubility Information Solubility in water: insoluble
Formula Weight 298.29
Percent Purity 94%
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl azodicarboxylate, 94%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dibenzyl azodicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'