missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dibenzothiophene-4-carboxaldehyde, 98%

Brand:  Alfa Aesar™ H64515.03

Code : NEW

Additional Details : CAS Number : 23985-81-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491759

  • £41.87 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Dibenzothiophene-4-carboxaldehyde
212.27
1567946
23985-81-1
MFCD00961967

Specifications

Dibenzothiophene-4-carboxaldehyde
Powder
Pale yellow
Air Sensitive
MFCD00961967
23985-81-1
98%
1g
212.27
1567946
Videos
SDS