missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzofuran, 98%, Thermo Scientific Chemicals
25.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.
Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.
Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 78°C to 83°C |
Density | 1.3 |
Boiling Point | 287°C to 289°C |
Flash Point | 130°C (266°F) |
Quantity | 25 g |
UN Number | UN3077 |
Beilstein | 121100 |
Solubility Information | Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents. |
Formula Weight | 168.2 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'