missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15402319
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5397-82-0
C13H8O2
196.21
MFCD00136272
OVJMIWIVPWPZMN-UHFFFAOYSA-N
dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
220843
O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
This item is not returnable. View return policy
5397-82-0
C13H8O2
196.21
MFCD00136272
OVJMIWIVPWPZMN-UHFFFAOYSA-N
dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
220843
O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1

Used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an pharmaceutical intermediate.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 74°C
Color White
Quantity 1 g
Beilstein 141044
Sensitivity Air Sensitive
IUPAC Name dibenzofuran-2-carbaldehyde
Formula Weight 196.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dibenzofuran-2-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'