missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ Diaphragm Valve

Hand operated diaphragm valves for RE 2.5/RZ 2.5/RE 6/RZ 6

£374.00 - £640.00

Specifications

Quantity 1 Each
Type Diaphragm Valve
View More Specs

Products 2
Product Code Brand Description For Use With Price Quantity & Availability  
Product Code Brand Description For Use With Price Quantity & Availability  
10273031
View Documents Promotion Details
VACUUBRAND™
20664010
Diameter: 6mm For RE 2.5/RZ 2.5/RE 6/RZ 6
£374.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
 
10120941
View Documents Promotion Details
VACUUBRAND™
20664011
Diameter: 16mm For RE 9/RZ 9/RE 16/RZ 16
£640.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
 
Description

Description

VM series valves are hand-operated diaphragm valves. They are used, e.g., as isolation valves, air-admittance valves or manual-control valves. By rotation of the wheel a flexible PTfE diaphragm is moved and pressed against the stainless steel seating or removed from it, closing and opening the valve, respectively. The gas-contacting parts are made of stainless steel and PTfE offering a good resistance to corrosive gases and vapors.
Specifications

Specifications

1 Each
Diaphragm Valve
Videos
SDS
Documents

Documents

Product Certifications