missing translation for 'onlineSavingsMsg'
Learn More

Diallyl phthalate, 98%, Thermo Scientific Chemicals

Catalog Number 10559453
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
131-17-9
C14H14O4
246.26
MFCD00008646
QUDWYFHPNIMBFC-UHFFFAOYSA-N
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
8560
bis(prop-2-enyl) benzene-1,2-dicarboxylate
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
This item is not returnable. View return policy
131-17-9
C14H14O4
246.26
MFCD00008646
QUDWYFHPNIMBFC-UHFFFAOYSA-N
diallyl phthalate, allyl phthalate, diallylphthalate, dapon r, phthalic acid diallyl ester, dapon 35, phthalic acid, diallyl ester, o-phthalic acid, diallyl ester, 1,2-benzenedicarboxylic acid, di-2-propenyl ester, unii-f79l0ul6st
8560
bis(prop-2-enyl) benzene-1,2-dicarboxylate
C=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70.0°C
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 300.0°C
Flash Point 166°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5180 to 1.5200
Quantity 250 mL
UN Number 3082
Beilstein 09, 4120
Specific Gravity 1.12
Solubility Information Solubility in water: 148mg/l (20°C)
Viscosity 13 mPa.s (20°C)
Formula Weight 246.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diallyl phthalate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diallyl phthalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'