Learn More
Alfa Aesar™ Diaion™ WK40L(H), ion exchange resin, weakly acidic porous type, 4.4 meq/ml on polyacrylate
Brand:  Alfa Aesar™ 046797.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity 1kg
Product Code. 15484085
  • £73.90 / 1kg
Estimated Shipment date
from Supplier 31-05-2018
Add to basket
Description
Description
  • Pharmaceuticals, dealkalination, metal recovery, antibiotics, sucrose purifications
  • Can be used in wide pH range
Specifications
Specifications
Diaion™ WK40L (H), ion exchange resin
425 to 1180μm
1kg
Weakly acidic porous type, 4.4meq/mL on polyacrylate
Beads