missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ WA21J, ion exchange resin, weakly basic porous type, 2.0 meq/ml on poly(styrene-divinylbenzene), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Deacidification, solvent treatment, refining of gelatin, adsorption of aldehydes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | 300-1180μm Beads |
CAS | 65945-33-7 |
Odor | Amine-like |
Particle Size | 300 to 1180μm |
Molecular Formula | C23H31N3 |
Color | Yellow |
Synonym | 1,2-ethanediamine, n-2-aminoethyl-n-ethenylphenyl methyl-, polymer with diethenylbenzene, 1,2-ethanediamine, n1-2-aminoethyl-n1-ethenylphenyl methyl-, polymer with diethenylbenzene, bis 2-aminoethyl 2-ethenylphenyl methyl amine; divinylbenzene, n'-2-aminoethyl-n'-2-ethenylphenyl methyl ethane-1,2-diamine; 1,2-bis ethenyl benzene |
Quantity | 1 kg |
InChI Key | UUZFYSHIFAAYRA-UHFFFAOYSA-N |
SMILES | C=CC1=CC=CC=C1C=C.NCCN(CCN)CC1=CC=CC=C1C=C |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'