missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Diaion™ UBK555(Ca), ion exchange fractionation resin, 8% x-linked, strongly acidic gel type, 2.0meq/mL on PS-DVB
Brand: Alfa Aesar™ 046731.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity | 1kg |
---|
Description
- Chromatographic sugar fractionation , fructose/glucose separations, sugar alcohol separations
Specifications
Diaion™ UBK555 (Ca), ion exchange fractionation resin | |
Odorless | |
Beads | |
1kg |
8% cross-linked, strongly acidic gel type, 2.0meq/mL on PS-DVB | |
220μm | |
Brown |