Learn More
Alfa Aesar™ Diaion™ SK112(Na), ion exchange resin, 12% cross-linked, strongly acidic gel type, 2.1 meq/ml on PS-DVB
Brand:  Alfa Aesar™ 046976.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity 1kg
Product Code. 15474095
  • £51.10 / 1kg
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Description
Description
  • Water softening, metal recovery, dehydration of solvents, amino acid purification
  • High resistance to oxidative attack
Specifications
Specifications
Diaion™ SK112 (Na), ion exchange resin
300 to 1180μm
1kg
12% cross-linked, strongly acidic gel type, 2.1meq/mL on PS-DVB
Beads