missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ PK228(Na), ion exchange resin, 14% cross-linked, strongly acidic porous type, 2.05 meq/ml on PS-DVB, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation. Used in solutions of high electrolyte content
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
- Used in solutions of high electrolyte content
Specifications
Physical Form | 300-1180μm Beads |
CAS | 63182-08-1 |
Odor | Odorless |
Particle Size | 300 to 1180μm |
Molecular Formula | C18H19NaO3S |
MDL Number | MFCD00132707 |
Color | Brown |
Synonym | amberjet r imac hp1110 |
Quantity | 1 kg |
InChI Key | MRTSCADIYLSZEU-UHFFFAOYSA-M |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'