missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diaion™ PK216(Na), ion exchange resin, 8% cross-linked, strongly acidic porous type, 1.75 meq/ml on PS-DVB

Brand:  Alfa Aesar™ 046718.A1

Code : NEW

Additional Details : Weight : 1.00000kg

Quantity 1kg

Product Code. 15484075

  • £56.10 / 1kg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

  • Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
Specifications

Specifications

Diaion™ PK216 (Na), ion exchange resin
Odorless
Beads
8% cross-linked, strongly acidic porous type, 1.75meq/mL on PS-DVB
300 to 1180μm
1kg
Videos
SDS