Learn More
Diaion™ HPA25(Cl), ion exchange resin, highly cross-linked, strongly basic highly porous type I, 0.5 meq/ml on PS-DVB, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 046691.A1
Code : NEW
Additional Details : CAS Number : 82643-04-7 Weight : 1.00000kg
Description
Enzyme immobilization, catalysis, used in high oxidative environments
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
250μm Beads | |
Highly cross-linked, strongly basic highly porous type I, 0.5meq/mL on PS-DVB | |
250μm |
Diaion™ HPA25 (Cl), ion exchange resin | |
82643-04-7 | |
1 kg |
RUO – Research Use Only