missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diaion™ HP21, synthetic adsorbent resin, highly porous type, 40g/L on PS-DVB, P.R. 110 angstroms, Thermo Scientific™
Click to view available options
Quantity:
1 kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Physical Form | 250μm Beads,S.A. 640m |
CAS | 9052-95-3 |
Odor | Odorless |
Particle Size | 250μm, P.R. 110Å |
Molecular Formula | C28H30 |
MDL Number | MFCD00148061 |
Density | 1.05 g/mL |
Color | White to Yellow |
Synonym | porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated |
Quantity | 1 kg |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'