Learn More
Alfa Aesar™ Diaion™ CR11(Na), chelating resin, highly porous type, 0.5 mmole/ml on poly(styrene-divinylbenzene)
Hardness removal in brine, selective transitional metal removal
Brand:  Alfa Aesar™ 046544.A1
Code : NEW
Additional Details : Weight : 1.00000kg
Quantity 1kg
Product Code. 15404055
  • £83.40 / 1kg
Estimated Shipment date
from Supplier 04-05-2018
Add to basket
Specifications
Specifications
Diaion™ CR11 (Na), chelating resin
Odorless
Beads
Highly porous type, 0.5mmole/mL on poly(styrene-divinylbenzene)
355 to 1180μm
1kg