missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-Butylchlorosilane, 95%, ACROS Organics™

Brand:  Acros Organics 432900050

Code : 29

Additional Details : CAS Number : 56310-18-0 Weight : 0.05500kg Transport : UN number : 2986 Chem class : 1a Pack group : II


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 11925921

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Di-tert-Butylchlorosilane
95%
178.78
≥94%
C8H19ClSi

Specifications

Di-tert-Butylchlorosilane
Authentic
1.4400 to 1.4420 (20°C, 589nm)
≥94%
C8H19ClSi
0.8840g/mL
5g
56310-18-0
Glass bottle
Liquid
95%
178.78
Clear colorless
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Di-tert-butylchlorosilane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H261-In contact with water releases flammable gases.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS