missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-tert-butyl iminodicarboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and air.
Di-tert-butyl-iminodicarboxylate may be used in the preparation of trans-diamino-2-butene and cis-1,4-diamino-2-butene; di-tert-butyl N-3-butenyliminodicarboxylate; N,N-di-tert-butyl[(2-fluoro-4-nitro)benzylamino]dicarboxylate & C1-C20 and C21-C40 fragments of tetrafibricin. Modified Gabriel Reagent.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and air.
Specifications
Melting Point | 114°C to 119°C |
Beilstein | 1911172 |
Solubility Information | Insoluble in water. |
Formula Weight | 217.27 |
Percent Purity | 98% |
Quantity | 1 g |
Sensitivity | Light sensitive |
Chemical Name or Material | Di-tert-butyl iminodicarboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'