Learn More
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 426808000
189.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 24424-99-5 Weight : 0.73040kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Di-tert-butyl dicarbonate | |
24424-99-5,109-99-9 | |
0.9130g/mL | |
AcroSeal™ Glass Bottle | |
O[CO2C(CH3)3]2 | |
800 mL | |
15, 3038 | |
DYHSDKLCOJIUFX-UHFFFAOYSA-N | |
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate | |
90495 | |
218.25 |
1M Solution in THF | |
Colorless | |
−17°C | |
C10H18O5 | |
MFCD00008805 | |
0.913 | |
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate | |
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C | |
218.25 | |
CHEBI:48500 | |
Liquid |
Safety and Handling
- Di-tert-butyl dicarbonate, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only