missing translation for 'onlineSavingsMsg'
Learn More

Di-tert-butyl dicarbonate, 97%, Thermo Scientific Chemicals

Catalog Number 10763191
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
24424-99-5
C10H18O5
218.25
MFCD00008805
DYHSDKLCOJIUFX-UHFFFAOYSA-N
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
90495
CHEBI:48500
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
24424-99-5
C10H18O5
218.25
MFCD00008805
DYHSDKLCOJIUFX-UHFFFAOYSA-N
di-tert-butyl dicarbonate, boc anhydride, boc-anhydride, di-t-butyl dicarbonate, di-tert-butyl pyrocarbonate, bis tert-butoxycarbonyl oxide, di-tert-butyldicarbonate, boc2o, di-t-butyl pyrocarbonate, di tert-butyl carbonate
90495
CHEBI:48500
tert-butyl (2-methylpropan-2-yl)oxycarbonyl carbonate
CC(C)(C)OC(=O)OC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 22.0°C to 24.0°C
Color White
Density 1.0200g/mL
Boiling Point 56.0°C to 57.0°C (0.5 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula O[CO2C(CH3)3]2
Refractive Index 1.4075 to 1.4095
Quantity 100 g
UN Number 2930
Fieser 04,128; 07,91; 08,145; 10,122; 12,159; 13,94; 15,113
Merck Index 15, 3038
Specific Gravity 1.02
Formula Weight 218.25
Percent Purity 97%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material Di-tert-butyl Dicarbonate, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-tert-butyl dicarbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'